Learn More
Medchemexpress LLC HY-111444 5mg Medchemexpress, Auxinole CAS:86445-22-9 Purity:>98%
SDP
Catalog No. 501968869
Encompass_Preferred
Medchemexpress, HY-111444 5mg Auxinole CAS:86445-22-9 Auxinole is a potent auxin antagonist of TIR1/AFB receptors, binding TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and so inhibits auxin-responsive gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8869 Supplier Medchemexpress LLC Supplier No. HY1114445MG
Medchemexpress, HY-111444 5mg Auxinole CAS:86445-22-9 Auxinole is a potent auxin antagonist of TIR1/AFB receptors, binding TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and so inhibits auxin-responsive gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.