Learn More
Gold Biotechnology Inc Auxo Agro EHA105 Electro Cells
SDP
Catalog No. 5000562304
Encompass_Preferred
GoldBio s Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency
Catalog No. 50-005-62304 Supplier Gold Biotechnology Inc Supplier No. CC26815X50
GoldBio s Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency