Learn More
Gold Biotechnology Inc Auxo Agro LBA4404 Comp Cells
SDP
Catalog No. 5000562214
Encompass_Preferred
GoldBio s new Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency
Catalog No. 50-005-62214 Supplier Gold Biotechnology Inc Supplier No. CC16715X50
GoldBio s new Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency