Learn More
Gold Biotechnology Inc Auxo Agro LBA4404 Electro
SDP
Catalog No. 5000562213
Encompass_Preferred
GoldBio s Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency
Catalog No. 50-005-62213 Supplier Gold Biotechnology Inc Supplier No. CC26710X50
GoldBio s Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency