Learn More
Apexbio Technology LLC AUY922 (NVP-AUY922), 100mg. Cas: 747412-49-3 MFCD: MFCD14635361. Potent Hsp90 inhibitor
SDP
Supplier:  Apexbio Technology LLC A4057100
Encompass_Preferred
AUY922 (NVP-AUY922) is a potent, novel synthetic resorcinylic isoxazole amide inhibitor of heat shock protein 90 (HSP90).[1] It is a small molecular with the formula of C26H31N3O5 and molecular weight of 465.5. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4869