Learn More
Apexbio Technology LLC AUY922 (NVP-AUY922) 747412-49-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064222
Encompass_Preferred
AUY922 (NVP-AUY922 CAS 747412-49-3) is a synthetic small molecule inhibitor belonging to the resorcinylic isoxazole amide class that selectively targets heat shock protein 90 (HSP90) AUY922 specifically interacts with the ATP-binding site of the HSP90 N-terminal domain disrupting its chaperone-dependent stabilization of client proteins involved in cell proliferation and survival It demonstrates antiproliferative activity against human gastric cancer cells with reported GI50 values ranging approximately from 2 to 40 nmol/L promoting the proteasomal degradation of receptor tyrosine kinases and other relevant client proteins This compound is widely used in preclinical research settings investigating HSP90-dependent signaling pathways in cancer
Catalog No. 50-000-64222 Supplier Apexbio Technology LLC Supplier No. A40575.1
AUY922 (NVP-AUY922 CAS 747412-49-3) is a synthetic small molecule inhibitor belonging to the resorcinylic isoxazole amide class that selectively targets heat shock protein 90 (HSP90) AUY922 specifically interacts with the ATP-binding site of the HSP90 N-terminal domain disrupting its chaperone-dependent stabilization of client proteins involved in cell proliferation and survival It demonstrates antiproliferative activity against human gastric cancer cells with reported GI50 values ranging approximately from 2 to 40 nmol/L promoting the proteasomal degradation of receptor tyrosine kinases and other relevant client proteins This compound is widely used in preclinical research settings investigating HSP90-dependent signaling pathways in cancer