Learn More
Selleck Chemical LLC Luminespib (NVP-AUY922) 5mg 747412-49-3 AUY-922
SDP
Catalog No. 508125
Encompass_Preferred
Luminespib (AUY-922, NVP-AUY922) is a highly potent HSP90 inhibitor for HSP90?/? with IC50 of 13 nM /21 nM in cell-free assays, weaker potency against the HSP90 family members GRP94 and TRAP-1, exhibits the tightest binding of any small-molecule HSP90 ligand. Luminespib (AUY-922, NVP-AUY922) effectively downregulates and destabilizes the IGF-1R? protein and results in growth inhibition, autophagy and apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-812-5 Supplier Selleck Chemical LLC Supplier No. S10695MG
Luminespib (AUY-922, NVP-AUY922) is a highly potent HSP90 inhibitor for HSP90?/? with IC50 of 13 nM /21 nM in cell-free assays, weaker potency against the HSP90 family members GRP94 and TRAP-1, exhibits the tightest binding of any small-molecule HSP90 ligand. Luminespib (AUY-922, NVP-AUY922) effectively downregulates and destabilizes the IGF-1R? protein and results in growth inhibition, autophagy and apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.