Learn More
TARGETMOL CHEMICALS INC AUZ 454 10MG
SDP
Catalog No. 502576430
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AUZ 454 (K03861) is a type II CDK2 inhibitor with Kd of 50 nM 18.6 nM 15.4 nM and 9.7 nM for CDK2(WT) CDK2(C118L) CDK2(A144C) and CDK2(C118L/A144C) respectlvely. purity: 99%
Catalog No. 50-257-6430 Supplier TARGETMOL CHEMICALS INC Supplier No. T686610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AUZ 454 (K03861) is a type II CDK2 inhibitor with Kd of 50 nM 18.6 nM 15.4 nM and 9.7 nM for CDK2(WT) CDK2(C118L) CDK2(A144C) and CDK2(C118L/A144C) respectlvely. purity: 99%