Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ AV-2000™ Facepiece

Catalog No. SCT80419102
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
SCT80419102 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCT80419102 Supplier Scott Safety™ Supplier No. 80419102

Interchangeable with Scott's full line of air-supplied, air-purifying, and communications products

Ideal for firefighting, industrial, and hazmat service. Available in four color-coded sizes to effectively fit those with different facial configurations.

  • Cone-shaped lens provides widest possible field of vision and helps reduce carbon dioxide buildup
  • Easily disassembled for cleaning and inspection
  • Comfortable, easy-to-remove nosecup
  • Double lip design provides positive pressure to the seal
  • Broad, deep chin cup allows great contact
  • Comfort Seal™ model features sealing edge with wide, soft surface for comfort

Certifications

NIOSH approved.

Specifications

Certifications/Compliance NIOSH
Color Black
Material Dupont™ Kevlar™ aramid fiber, EPDM Rubber Seal
Product Type Respiratory Facepiece
Format Large
For Use With (Application) Ideal for firefighting, industrial, hazmat service
For Use With (Equipment) Scott air supplied, air purifying, communication products
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.