Learn More
Scott Safety™ AV-3000 HT Facepiece, Approved to NFPA 1981™ 2013 EDITION STANDARD
Catalog No. 19222929
Change view
Click to view available options
Size:
Medium
Catalog No. Size
19-222-929 Medium
1 options
Catalog No. 19-222-929 Supplier Scott Safety™ Supplier No. 7012472117
Please contact your local Fisher Scientific sales representative for pricing
Uses proven SureSeal faceseal technology for comfort, fit and performance.
Scott Safety's AV-3000 HT facepiece meets all requirements established in the NFPA 1981:2013 Edition standard, with state-of-the-art material technology to provide enhanced thermal durability and voice intelligibility performance for added safety.
  • Approved with 2013 Edition Air-Pak X3 and Air-Pak 75 SCBAs
  • Approved with NFPA 1981, 2002 and 2007 Edition Air-Pak SCBAs
  • State-of-the-art materials and design for enhanced performance
  • ANSI Z87.1-2010 Impact Rated
  • Compatible with EPIC and EPIC 3 Communication Systems
  • Right and Left communication installation options
  • Designed for Top Down Convertibility (CBRN APR approval pending)
  • Compatible with Scott Sight In-Mask Thermal Imager
Features
  • Meets ANSI Z87.1-2010 standard for impact rating and shall be marked as Z87+
  • Smaller, Kevlar head harness eliminates bunching and discomfort under helmets
  • High performance lens and bezel for improved thermal performance
  • Lens is a single, replaceable, modified cone configuration constructed of high temperature and radiant heat resistant non-shatter type polycarbonate material
  • High visibility inhalation valves allow for easy inspection
  • Dual voice emitters for enhanced voice transmission around the wearer
  • Positioning tab assists with proper and quick donning
Certifications
NIOSH approved
Specifications
Certifications/Compliance ANSI Z87.1-2010
Color Black
Product Type Facepiece
Harness Style 4-strap
Material Kevlar™ aramid fiber, Polycarbonate Lens
Model AV-3000 HT
Size Medium