Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ AV-3000™ HT Facepiece with Aramid Fibers Head Harness

Catalog No. p-4621393
Change view
Click to view available options
Certifications/Compliance:
ANSI Z87.1-2010, NFPA 1981, 2002, 2007
ANSI Z87.1-2010, NFPA 1981, 2002, 2007, 2013
NFPA 1981, 2002, 2007, 2013
Includes:
Facepiece and harness only
Left side mounting communication bracket
Right side mounting communication bracket
Size:
Small
Medium
Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Includes Size
19-100-850 NFPA 1981, 2002, 2007, 2013 Facepiece and harness only Small
19-100-851 NFPA 1981, 2002, 2007, 2013 Facepiece and harness only Medium
19-100-852 NFPA 1981, 2002, 2007, 2013 Facepiece and harness only Large
19-100-856 ANSI Z87.1-2010, NFPA 1981, 2002, 2007, 2013 Left side mounting communication bracket Small
19-100-857 ANSI Z87.1-2010, NFPA 1981, 2002, 2007, 2013 Left side mounting communication bracket Medium
19-100-858 ANSI Z87.1-2010, NFPA 1981, 2002, 2007, 2013 Left side mounting communication bracket Large
19-100-853 ANSI Z87.1-2010, NFPA 1981, 2002, 2007, 2013 Right side mounting communication bracket Small
19-100-854 ANSI Z87.1-2010, NFPA 1981, 2002, 2007, 2013 Right side mounting communication bracket Medium
19-100-855 ANSI Z87.1-2010, NFPA 1981, 2002, 2007 Right side mounting communication bracket Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-100-850 Supplier Scott Safety™ Supplier No. 7012511832

Please contact your local Fisher Scientific sales representative for pricing

Utilizes the proven Sureseal™ technology – a reverse reflex sealing surface that maintains a continuous seal around the user's face to provide enhanced comfort and reliable fit.

  • Proprietary lens and bezel material to enhance thermal durability and wearer safety
  • Redesigned voice communication components to deliver improved intelligibility with or without communication devices
  • Robust communication bracket attachment with right- or left-side mounting for user preference
  • Compatible with current and new EPIC 3 communication devices
  • Designed for Top-Down Convertibility, the AV-3000 HT will allow for a single facepiece to be used for all NFPA™/CBRN/NIOSH† respiratory applications, reducing fit testing, training and inventory for lower total cost of ownership (pending approvals)
  • Approved with NFPA 1981, 2002 and 2007 Edition Air-Pak SCBAs
  • ANSI™ Z87.1-2010 Impact Rated

NFPA 1981 2013 Edition Facepiece Requirements
The AV-3000 HT is designed to meet the facepiece requirements set forth in the 2013 Edition of the NFPA 1981 Standard for high heat and flame and radiant heat by utilizing new proprietary lens and bezel materials. These new materials provide enhanced thermal durability and enhanced wearer safety by ensuring the facepiece stays under positive pressure when exposed to the heat and flame loads measured under NFPA 2013.
NFPA 1981 2013 Edition Voice Intelligibility Tests
Two voice intelligibility tests' one for mechanical communications and one for electronic, or amplified, communications' are included in the NFPA 2013 Edition.
The AV-3000 HT:

  • Meets the mechanical and electronic testing requirements set forth in the new standard
  • Uses two voicemitters to provide a sphere of intelligibility to those around the respirator wearer, not just the individuals directly in front of the wearer
  • Enables right- or left-side mounting for Scott Safety EPIC 3 voice amplifiers

†AV-3000 HT:

  • Is Approved for use with the 2002 and 2007 editions of the Scott Safety Air-Pak Fifty, AP75, NxG2 and NxG7 respirators
  • CANNOT be approved on '97 Air-Pak

Specifications

Certifications/Compliance NFPA 1981, 2002, 2007, 2013
Color Black
Product Type Facepiece
For Use With (Application) Firefighting
For Use With (Equipment) Air-Pak™ X3 Pro SCBA, EPIC 3 Communication Devices
Includes Facepiece and harness only
Material Dupont™ Kevlar™ aramid fiber
Size Small
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.