Learn More
Medchemexpress LLC HY-10346 5mg Medchemexpress, AV-412 CAS:451493-31-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103465MG
Encompass_Preferred
Medchemexpress, HY-10346 5mg AV-412 CAS:451493-31-5 AV-412 (MP412) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8111