Learn More
Selleck Chemical LLC AV-412 free base S2733-2mg
SDP
Catalog No. 503111834
Encompass_Preferred
AV-412 free base (MP-412 free base) is an EGFR inhibitor with IC50s of 0 75 0 51 0 79 2 30 19 nM for EGFR EGFRL858R EGFRT790M EGFRL858R/T790M and ErbB2 respectively
Catalog No. 50-311-1834 Supplier Selleck Chemical LLC Supplier No. S27332MG
AV-412 free base (MP-412 free base) is an EGFR inhibitor with IC50s of 0 75 0 51 0 79 2 30 19 nM for EGFR EGFRL858R EGFRT790M EGFRL858R/T790M and ErbB2 respectively