Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scott™ AV-632 Bayonet Adapter

Catalog No. 19072330
Change view
Click to view available options
Quantity:
5 adapters/box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-330 5 adapters/box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-072-330 Supplier 3M™ Scott™ Supplier No. 7100378707

Please contact your local Fisher Scientific sales representative for pricing

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to be used with AV-3000 facepieces and 3M™ cartridges 6000 series, filter 7093 series and filters 2000 series.

  • For use with full facepiece respirators AV-3000 with SureSeal and AV-3000 HT
  • Adapter allows bayonet cartridges and filters to be used with full facepiece respirators
  • Includes 5 adapters in a box
  • NIOSH approved with AV-3000 with SureSeal and AV-3000 HT full facepiece respirators
  • Simple, quick quarter turn for easy attachment

Specifications

Certifications/Compliance NIOSH
Connection Type Bayonet
Color Black
Product Type Adapter
For Use With (Equipment) AV-3000 HT, AV-3000 SureSeal Facepieces, 3M™ bayonet cartridges and filters
Model AV-632
Quantity 5 adapters/box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.