Description
- AVA easily automates chemistry procedures
- Makes chemistry safer by running unattended experiments safely, reducing manual errors, setting apparatus safety limits to prevent accidents
- Configures audio and visual alarms
- Advantages of software automation include improved productivity, and easy results sharing between users which improves research/collaboration
- AVA software can:
- Control, log common laboratory apparatus and processes
- Eliminate time-consuming, manual data recording
- Track what you do, as you do it and repeat in the experiment scheduler
- Visualize results for rapid analysis and reporting
- Link devices for dual control (pump and balance for additions)
- Control up to four experiments in parallel
Specifications
Specifications
| Type | Level 3 |
| Includes | Controller, Data Hub, and AVA Installed |
| Product Line | AVA |