Learn More
Heidolph™ AVA Software Kit - US Plug
SDP
Catalog No. 13876721 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Level 4
Catalog No. Type
13-876-721 Level 4
1 options
Catalog No. 13-876-721 Supplier Heidolph™ Supplier No. 015200008
Add to Cart
Add to Cart
Request a Quote
Heidolph™ AVA Software Kit - US Plug includes Controller, Data Hub, and AVA Installed
  • AVA easily automates chemistry procedures
  • Makes chemistry safer by running unattended experiments safely, reducing manual errors, setting apparatus safety limits to prevent accidents
  • Configures audio and visual alarms
  • Advantages of software automation include improved productivity, and easy results sharing between users which improves research/collaboration
  • AVA software can:
    • Control, log common laboratory apparatus and processes
    • Eliminate time-consuming, manual data recording
    • Track what you do, as you do it and repeat in the experiment scheduler
    • Visualize results for rapid analysis and reporting
    • Link devices for dual control (pump and balance for additions)
    • Control up to four experiments in parallel
Specifications
Type Level 4
Includes Controller, Data Hub, and AVA Installed
Product Line AVA
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov