Learn More
TARGETMOL CHEMICALS INC AVACOPAN 5MG
SDP
Catalog No. 502579089
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avacopan (CCX168) is an orally administered small-molecule C5aR ( IC50 0.1 nM.)antagonist that blocks the effects of C5a and prevents the development of GN induced by anti-myeloperoxidase antibodies in a murine model of AAV. purity: 99%
Catalog No. 50-257-9089 Supplier TARGETMOL CHEMICALS INC Supplier No. T82235MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avacopan (CCX168) is an orally administered small-molecule C5aR ( IC50 0.1 nM.)antagonist that blocks the effects of C5a and prevents the development of GN induced by anti-myeloperoxidase antibodies in a murine model of AAV. purity: 99%