Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Avagard™ D Instant Hand Antiseptic with Moisturizers (61% w/w ethyl alcohol) 9222

Catalog No. 19072386
Encompass
Change view
Click to view available options
Volume (English):
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-072-386 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-386 Supplier 3M™ Supplier No. 7000002748
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avagard™ D Instant Hand Antiseptic provides fast and effective bacterial kill without being hard on skin.

  • Broad-spectrum activity kills harmful bacteria, including antibiotic-resistant strains (in vitro)
  • Proven efficacy - meets CDC, AORN and WHO Guidelines
  • Helps prevent skin breakdown.
  • Dries quickly without leaving hands sticky
  • Compatible with CHG
  • Recommended for repeat use
  • Reference: 1APIC Guideline for Handwashing and Hand Antisepsis in Health Care Settings, 1995. Larson, E.L. 2CDC Guideline for Handwashing and Hospital Environmental Control, 1985. Julia S. Garner; Martin S. Favero, Hospital Infections Program Center for Infectious Diseases, Centers for Disease Control and Prevention
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Composition 61% w/w ethyl alcohol
Volume (English) 16 oz.
Volume (Metric) 473 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.