Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Avagard™ D Instant Hand Antiseptic with Moisturizers (61% w/w ethyl alcohol) 9230

Catalog No. 19072388
Encompass
Encompass_Preferred
Change view
Click to view available options
Volume (English):
33.8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-072-388 33.8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-072-388 Supplier 3M™ Supplier No. 7100073223
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Avagard™ D Instant Hand Antiseptic provides fast and effective bacterial kill without being hard on skin.

  • Broad-spectrum activity kills harmful bacteria, including antibiotic-resistant strains (in vitro)
  • Proven efficacy - meets CDC, AORN and WHO Guidelines
  • Helps prevent skin breakdown.
  • Dries quickly without leaving hands sticky
  • Compatible with CHG
  • Recommended for repeat use
  • Reference: 1APIC Guideline for Handwashing and Hand Antisepsis in Health Care Settings, 1995. Larson, E.L. 2CDC Guideline for Handwashing and Hospital Environmental Control, 1985. Julia S. Garner; Martin S. Favero, Hospital Infections Program Center for Infectious Diseases, Centers for Disease Control and Prevention
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Composition 61% w/w ethyl alcohol
Volume (English) 33.8 oz.
Volume (Metric) 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.