Learn More
New England Biolabs, Inc. AvaI – 10000 units
SDP
Catalog No. 50812041
Encompass
A restriction endonuclease that recognizes the sequence C^YCGR_G.
Catalog No. 50-812-041 Supplier New England Biolabs, Inc. Supplier No. R0152L
A restriction endonuclease that recognizes the sequence C^YCGR_G.