Learn More
New England Biolabs, Inc. AvaI – 2000 units
SDP
Catalog No. 50812043
Encompass
A restriction endonuclease that recognizes the sequence C^YCGR_G.
Catalog No. 50-812-043 Supplier New England Biolabs, Inc. Supplier No. R0152S
A restriction endonuclease that recognizes the sequence C^YCGR_G.