Learn More
New England Biolabs, Inc. AvaI – 10000 units
SDP
Supplier:  New England Biolabs, Inc. R0152M
Encompass
A restriction endonuclease that recognizes the sequence C^YCGR_G.
Catalog No. 50-812-042