Learn More
New England Biolabs, Inc. AvaII – 10000 units
SDP
Catalog No. 50812045
Encompass
A restriction endonuclease that recognizes the sequence G^GWC_C.
Catalog No. 50-812-045 Supplier New England Biolabs, Inc. Supplier No. R0153L
A restriction endonuclease that recognizes the sequence G^GWC_C.