Learn More
New England Biolabs, Inc. AvaII – 2000 units
SDP
Catalog No. 50812047
Encompass
A restriction endonuclease that recognizes the sequence G^GWC_C.
Catalog No. 50-812-047 Supplier New England Biolabs, Inc. Supplier No. R0153S
A restriction endonuclease that recognizes the sequence G^GWC_C.