Learn More
Selleck Chemical LLC Avanafil 50mg 330784-47-9
SDP
Supplier:  Selleck Chemical LLC S401950MG
Encompass_Preferred
Avanafil is a highly selective PDE5 inhibitor with IC50 of 5.2 nM, >121-fold selectivity over other PDEs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3546