Learn More
Pyramex™ Avanté™ Safety Eyewear
Catalog No. 19155956 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Gun Metal
Silver
Silver, Black
Lens Tint:
Amber
Blue
Blue, Mirror
Clear
Gray
Mirror
Orange, Mirror
Silver, Mirror
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-956 Gun Metal Gray
19-155-727 Black Clear
19-155-728 Black Gray
19-155-730 Black Amber
19-155-731 Black Blue
19-155-732 Black Silver, Mirror
19-155-733 Black Blue, Mirror
19-155-734 Black Mirror
19-155-955 Gun Metal Clear
19-155-989 Silver Clear
19-155-990 Silver Gray
19-155-991 Silver Orange, Mirror
19-155-992 Silver Mirror
19-155-993 Silver Blue, Mirror
19-155-994 Silver, Black Clear
19-155-995 Silver, Black Gray
19-155-996 Silver, Black Silver, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 19-155-956 Supplier Pyramex™ Supplier No. SGM4520D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight, sports styling, contours to the face to provide maximum protection
  • Straight back flexible temples
  • Vented frame increases air flow to prevent fogging
  • Built-in rubber nose pads provide comfort fit and protect against slipping
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Gray
    Frame Color Gun Metal
    Lens Material Polycarbonate
    Frame Material Polycarbonate
    Frame Type 3 thin holes on the frame, one in the middle, one in left edge and onein the right edge