Learn More
Beckman Coulter Beckman Coulter Avanti J-15 Blood Pkg
Supplier: Beckman Coulter C19424
Blood Sample Package: Avanti J-15, Non-IVD, Ventilated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Bucket covers sold separately)