Learn More
Beckman Coulter Beckman Coulter Avanti J-15 Cell Culture Pkg
Catalog No. 501631951
Encompass_Preferred
Cell Culture Package: Avanti J-15, Non-IVD, Ventilated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Bucket covers sold separately)
This item is not returnable. View return policy
Catalog No. 50-163-1951 Supplier Beckman Coulter Supplier No. C19418
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Cell Culture Package: Avanti J-15, Non-IVD, Ventilated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Bucket covers sold separately)