Learn More
Beckman Coulter Beckman Coulter Avanti J-15R Blood Pkg
Catalog No. 501631946
Encompass_Preferred
Blood Sample Package: Avanti J-15R, Non-IVD, Refrigerated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Bucket covers sold separately)
This item is not returnable. View return policy
Catalog No. 50-163-1946 Supplier Beckman Coulter Supplier No. C19421
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Blood Sample Package: Avanti J-15R, Non-IVD, Refrigerated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 13 mm diameter tube adapters (qty 4) and 16 mm diameter tube adapters (qty 4). (Bucket covers sold separately)