Learn More
Beckman Coulter Beckman Coulter Avanti J-15R Cell Culture Pkg
Supplier:  Beckman Coulter C19415
Encompass_Preferred
Cell Culture Package: Avanti J-15R, Non-IVD, Refrigerated Benchtop Centrifuge, 100-120 V, 50/60 Hz, JS-4.750 (4 x 750 mL) swinging bucket rotor, 15 mL conical tube adapters (qty 4) and 50 mL conical tube adapters (qty 4). (Bucket covers sold separately)
Catalog No. 50-163-1944
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.