Learn More
Beckman Coulter Beckman Coulter Avanti J-15R refrigerated benchtop centrifuge, IVD, swinging-bucket package, 200-230V 50Hz
Catalog No. NC2178365
Encompass
Beckman Coulter Avanti J-15R is a high-performance refrigerated benchtop centrifuge designed for reliable sample separation in clinical and research laboratories. The swinging-bucket package provides flexible capacity and controls that minimize sample disturbance while supporting routine workflows.
  • High-speed performance up to 10,200 RPM for rapid separations.
  • Maximum relative centrifugal force of 11,420 xg to handle a range of sample types.
  • Temperature control from -10°C to +40°C in 1° increments to protect temperature-sensitive samples.
  • Swinging-bucket package includes a JS-4.750 rotor for flexible sample capacity.
  • Compact 55.6 cm (21.9 in) footprint saves benchtop space.
  • Intuitive color LCD and program memory for simplified operation.
This item is not returnable. View return policy
Catalog No. NC2178365 Supplier Beckman Coulter Supplier No. C19398
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Beckman Coulter Avanti J-15R is a high-performance refrigerated benchtop centrifuge designed for reliable sample separation in clinical and research laboratories. The swinging-bucket package provides flexible capacity and controls that minimize sample disturbance while supporting routine workflows.
  • High-speed performance up to 10,200 RPM for rapid separations.
  • Maximum relative centrifugal force of 11,420 xg to handle a range of sample types.
  • Temperature control from -10°C to +40°C in 1° increments to protect temperature-sensitive samples.
  • Swinging-bucket package includes a JS-4.750 rotor for flexible sample capacity.
  • Compact 55.6 cm (21.9 in) footprint saves benchtop space.
  • Intuitive color LCD and program memory for simplified operation.