Learn More
Beckman Coulter AVANTI JXN26 200-240V 50HZ
Catalog No. NC2056934
Encompass
  • High-speed refrigerated centrifuge for efficient sample processing
  • Switched Reluctance drive system for powerful operation
  • Temperature range of -10°C to +40°C in 1° increments
  • Maximum speed of 26,000 rpm for high-throughput applications
  • Maximum capacity of 6.0 L
  • User-defined programs capability up to 1,000
  • Automatic rotor identification for enhanced safety
  • Noise output levels less than 62 dBA
  • Built-in safety features including BioSafe options and Dynamic Rotor Inertia Check
  • Ergonomically designed for ease of use
This item is not returnable. View return policy
Catalog No. NC2056934 Supplier Beckman Coulter Supplier No. B34183
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
  • High-speed refrigerated centrifuge for efficient sample processing
  • Switched Reluctance drive system for powerful operation
  • Temperature range of -10°C to +40°C in 1° increments
  • Maximum speed of 26,000 rpm for high-throughput applications
  • Maximum capacity of 6.0 L
  • User-defined programs capability up to 1,000
  • Automatic rotor identification for enhanced safety
  • Noise output levels less than 62 dBA
  • Built-in safety features including BioSafe options and Dynamic Rotor Inertia Check
  • Ergonomically designed for ease of use