Learn More
Avanti Polar Lipids Sphingomyelin (Milk, Bovine), 475662-40-9, MFCD00132361, 25 mg
Supplier:  Avanti Polar Lipids 860063P25MG
Encompass
Synonym: N-(tricosanoyl)-sphing-4-enine-1-phosphocholine, Formula Weight: 785.034, Purity >99%. As a major constituent of cell membranes, sphingomyelin is found at particularly high concentrations in the membranes of nerve cells (in the myelin sheaths) and red blood cells.
Catalog No. NC1576874