Learn More
Avantik AVANTIK S200 UV LASER PRINTER
SDP
Catalog No. NC3732755
Encompass_Preferred
  • UV laser slide printer
  • Dual hopper design
  • Slide capacity of up to 200 slides
  • Printing speed of 3-5 seconds per slide
  • Printing resolution of 2500 DPI
  • Print permanence is resistant to reagents, H&E, IHC staining, and handling
  • Compatible with any standard microscope slides
  • Integrated touchscreen computer for user interface
  • Software features unlimited templates and drag-and-drop positioning
  • LIS integration for text and image upload
  • Dimensions of 16 in x 9 in x 12.6 in
  • Benchtop weight of 60 lbs
  • Works on 100-240V, 60Hz
  • Non-contact UV laser marking technology
  • Built-in computer; no external PC required
  • Equipped with built-in activated charcoal filter for air purification
Catalog No. NC3732755 Supplier Avantik Supplier No. EQ0673
Add to Cart
Add to Cart
Request a Quote
  • UV laser slide printer
  • Dual hopper design
  • Slide capacity of up to 200 slides
  • Printing speed of 3-5 seconds per slide
  • Printing resolution of 2500 DPI
  • Print permanence is resistant to reagents, H&E, IHC staining, and handling
  • Compatible with any standard microscope slides
  • Integrated touchscreen computer for user interface
  • Software features unlimited templates and drag-and-drop positioning
  • LIS integration for text and image upload
  • Dimensions of 16 in x 9 in x 12.6 in
  • Benchtop weight of 60 lbs
  • Works on 100-240V, 60Hz
  • Non-contact UV laser marking technology
  • Built-in computer; no external PC required
  • Equipped with built-in activated charcoal filter for air purification