Learn More
Mac-Mod AVANTOR ACE EXCEL SUPERC18 90
SDP
Catalog No. NC2397136
Encompass
Avantor ACE Excel SuperC18, 90 A, 2.1/3.0 x 5 mm UHPLC Guard Cartridges (3/pk)
Catalog No. NC2397136 Supplier Mac-Mod Supplier No. ACE1P11GD2U
Avantor ACE Excel SuperC18, 90 A, 2.1/3.0 x 5 mm UHPLC Guard Cartridges (3/pk)