Learn More
Avantor™ Glass Florisil SPE Columns
Catalog No. 14650451 Shop All Avantor Products
Change view
Click to view available options
Sorbent:
Florisil
Catalog No. Sorbent
14-650-451 Florisil
1 options
Catalog No. 14-650-451 Supplier Avantor™ Supplier No. 742007
Add to Cart
Add to Cart
For adsorption chromatography with media Mg2SiO3
  • 1000mg; 8mL column
  • Designed to fit popular processors and racks
Specifications
Base Material Activated Magnesium Silicate
Column Type SPE
For Use With (Application) Gas Chromatography
Sorbent Florisil