Learn More
TARGETMOL CHEMICALS INC AVAPRITINIB 5MG
SDP
Catalog No. 502579811
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avapritinib (BLU-285) is a dual inhibitor of the mutant KIT receptor (KIT D816V IC50 0.27 nM) and PDGFR(alpha) (PDGFR(alpha) D842V IC50 0.24 nM). purity: 99%
Catalog No. 50-257-9811 Supplier TARGETMOL CHEMICALS INC Supplier No. T51095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avapritinib (BLU-285) is a dual inhibitor of the mutant KIT receptor (KIT D816V IC50 0.27 nM) and PDGFR(alpha) (PDGFR(alpha) D842V IC50 0.24 nM). purity: 99%