Learn More
Medchemexpress LLC HY-101561 5mg Medchemexpress, Avapritinib CAS:1703793-34-3 Purity:>98%
SDP
Catalog No. 501932168
Encompass_Preferred
Medchemexpress, HY-101561 5mg Avapritinib CAS:1703793-34-3 Avapritinib (BLU-285) is a highly potent, selective, and orally bioavailable KIT and PDGFRA activation loop mutant kinases inhibitor with IC 50 s of 0.27 and 0.24 nM for KIT D816V and PDGFRA D842V, respectively. Avapritinib (BLU-285) binds the active conformation of the kinase and shows antitumor activity. Avapritinib (BLU-285) attenuates the transport function of both ABCB1 and ABCG2 [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2168 Supplier Medchemexpress LLC Supplier No. HY1015615MG
Medchemexpress, HY-101561 5mg Avapritinib CAS:1703793-34-3 Avapritinib (BLU-285) is a highly potent, selective, and orally bioavailable KIT and PDGFRA activation loop mutant kinases inhibitor with IC 50 s of 0.27 and 0.24 nM for KIT D816V and PDGFRA D842V, respectively. Avapritinib (BLU-285) binds the active conformation of the kinase and shows antitumor activity. Avapritinib (BLU-285) attenuates the transport function of both ABCB1 and ABCG2 [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.