Learn More
Selleck Chemical LLC Avapritinib
SDP
Catalog No. 5000000571
Encompass_Preferred
Avapritinib is a small molecule kinase inhibitor that potently inhibits PDGFR D842V mutant activity in vitro (IC50 0 5 nM) and PDGFR D842V autophosphorylation in the cellular setting (IC50 30 nM) also a potent inhibitor of the analogous Kit (c-Kit) mutation D816V in Kit (c-Kit) Exon 17 (IC50 0 5 nM)
Catalog No. 50-000-00571 Supplier Selleck Chemical LLC Supplier No. S85535MG
Avapritinib is a small molecule kinase inhibitor that potently inhibits PDGFR D842V mutant activity in vitro (IC50 0 5 nM) and PDGFR D842V autophosphorylation in the cellular setting (IC50 30 nM) also a potent inhibitor of the analogous Kit (c-Kit) mutation D816V in Kit (c-Kit) Exon 17 (IC50 0 5 nM)