Learn More
Medchemexpress LLC HY-13215 10mg Medchemexpress, Avasimibe CAS:166518-60-1 Purity:>98%
SDP
Catalog No. 502030263
Encompass_Preferred
Medchemexpress, HY-13215 10mg Avasimibe CAS:166518-60-1 Avasimibe is an oral inhibitor of acyl-Coenzyme A:cholesterol acyltransferase (ACAT) with IC50s of 24 and 9.2 µM for ACAT1 and ACAT2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0263 Supplier Medchemexpress LLC Supplier No. HY1321510MG
Medchemexpress, HY-13215 10mg Avasimibe CAS:166518-60-1 Avasimibe is an oral inhibitor of acyl-Coenzyme A:cholesterol acyltransferase (ACAT) with IC50s of 24 and 9.2 µM for ACAT1 and ACAT2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.