Learn More
Selleck Chemical LLC Avasimibe 10mg 166518-60-1 CI-1011
SDP
Supplier:  Selleck Chemical LLC S218710MG
Encompass_Preferred
Avasimibe (CI-1011) inhibits ACAT with IC50 of 3.3 ?M, also inhibits human P450 isoenzymes CYP2C9, CYP1A2 and CYP2C19 with IC50 of 2.9 ?M, 13.9 ?M and 26.5 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-665-8