Learn More
Medchemexpress LLC Avasopasem manganese | 435327-40-5 | 96.1% | 483.38 | 10 MG
SDP
Catalog No. 5000344503
Encompass_Preferred
Avasopasem manganese (GC4419; M-40419) is a potent superoxide dismutase mimetic that rapidly and specifically converts O2*- to hydrogen peroxide (H2O2), arresting the initiation of this cascade. It can be used for the research of severe oral mucositis (SOM) and cancer.
  • Potent superoxide dismutase mimetic
  • Rapidly and specifically converts O2*- to hydrogen peroxide (H2O2)
  • Arrests the initiation of the O2*- cascade
  • Can be used for research of severe oral mucositis (SOM)
  • Can be used for research of cancer
  • Intended to interrupt severe oral mucositis pathogenesis
  • Reduces the duration, incidence, and severity of radiation-induced oral mucositis
Catalog No. 50-003-44503 Supplier Medchemexpress LLC Supplier No. HY1091101MG
Avasopasem manganese (GC4419; M-40419) is a potent superoxide dismutase mimetic that rapidly and specifically converts O2*- to hydrogen peroxide (H2O2), arresting the initiation of this cascade. It can be used for the research of severe oral mucositis (SOM) and cancer.
  • Potent superoxide dismutase mimetic
  • Rapidly and specifically converts O2*- to hydrogen peroxide (H2O2)
  • Arrests the initiation of the O2*- cascade
  • Can be used for research of severe oral mucositis (SOM)
  • Can be used for research of cancer
  • Intended to interrupt severe oral mucositis pathogenesis
  • Reduces the duration, incidence, and severity of radiation-induced oral mucositis