Learn More
Cayman Chemical Avatrombopag, Purity: ≥98%, CAS: 570406-98-3, 10 mg
SDP
Catalog No. 502934648
Encompass_Preferred
A thrombopoietin receptor agonist; induces proliferation of Ba/F3 cells expressing the thrombopoietin receptor (EC50 = 3.3 nM); specific for human and chimpanzee; induces STAT5 phosphorylation in human and chimpanzee blood platelets; increases the number of human platelets in NOD/SCID mice transplanted with CD34+ human fetal liver cells at 1 and 3 mg/kg
Catalog No. 50-293-4648 Supplier Cayman Chemical Supplier No. 2653510 MG
A thrombopoietin receptor agonist; induces proliferation of Ba/F3 cells expressing the thrombopoietin receptor (EC50 = 3.3 nM); specific for human and chimpanzee; induces STAT5 phosphorylation in human and chimpanzee blood platelets; increases the number of human platelets in NOD/SCID mice transplanted with CD34+ human fetal liver cells at 1 and 3 mg/kg