Learn More
Selleck Chemical LLC Avatrombopag
SDP
Catalog No. 5000002914
Encompass_Preferred
Avatrombopag (AKR-501 E5501 YM477) is an orally administered small-molecule thrombopoietin receptor (c-Mpl) agonist which increases platelet number but not platelet activation
Catalog No. 50-000-02914 Supplier Selleck Chemical LLC Supplier No. S66242MG
Avatrombopag (AKR-501 E5501 YM477) is an orally administered small-molecule thrombopoietin receptor (c-Mpl) agonist which increases platelet number but not platelet activation