Learn More
TARGETMOL CHEMICALS INC AVE-0118 5MG
SDP
Catalog No. 502580677
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AVE-0118 is a potassium channel blocker and suppresses persistent atrial fibrillation. purity: 99%
Catalog No. 50-258-0677 Supplier TARGETMOL CHEMICALS INC Supplier No. T266895MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AVE-0118 is a potassium channel blocker and suppresses persistent atrial fibrillation. purity: 99%