Learn More
TARGETMOL CHEMICALS INC AVE5688 5MG
SDP
Catalog No. 502581724
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AVE5688 is an inhibitor of glycogen phosphorylase (GP IC50s 430 nM and 915 nM; Kds 170 nM and 530 nM for rmGPb and rmGPa). It can be used for the research of type 2 diabetes. purity: 100%
Catalog No. 50-258-1724 Supplier TARGETMOL CHEMICALS INC Supplier No. T104215MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AVE5688 is an inhibitor of glycogen phosphorylase (GP IC50s 430 nM and 915 nM; Kds 170 nM and 530 nM for rmGPb and rmGPa). It can be used for the research of type 2 diabetes. purity: 100%