Learn More
Medchemexpress LLC HY-108730 1mg Medchemexpress, Avelumab CAS:1537032-82-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1087301MG
Encompass_Preferred
Medchemexpress, HY-108730 1mg Avelumab CAS:1537032-82-8 Avelumab is a fully human IgG1 anti-PD-L1 monoclonal antibody with potential antibody-dependent cell-mediated cytotoxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8623