Learn More
Cayman Chemical Avenaciolide, Purity: ≥95%, CAS: 26057-70-5, 2.5 mg
SDP
Catalog No. 502937241
Encompass_Preferred
A fungal metabolite; inhibits glutamate transport in isolated rat liver mitochondria (Ki = 8 μM)
Catalog No. 50-293-7241 Supplier Cayman Chemical Supplier No. 281282.5 MG
A fungal metabolite; inhibits glutamate transport in isolated rat liver mitochondria (Ki = 8 μM)