Learn More
Medchemexpress LLC Avenanthramide D | 115610-36-1 | 99.9% | 283.28 | C16H13NO4 | 5 MG
SDP
Catalog No. 5000228968
Encompass_Preferred
Avenanthramide D is a phenolic acid found in oat (Avena sativa) that is used in biochemical and inflammation research. Supplied as a powder for in vitro assays and compound screening.
  • Phenolic acid researched for anti-inflammatory activity.
  • High purity suitable for biochemical assays.
  • Soluble in DMSO; ultrasonic assistance may be required.
  • Store protected from light at 4°C; in solution keep at -80°C for long-term storage.
  • Available in small research quantities, such as 5 mg, for assay development.
Catalog No. 50-002-28968 Supplier Medchemexpress LLC Supplier No. HY1186935MG
Avenanthramide D is a phenolic acid found in oat (Avena sativa) that is used in biochemical and inflammation research. Supplied as a powder for in vitro assays and compound screening.
  • Phenolic acid researched for anti-inflammatory activity.
  • High purity suitable for biochemical assays.
  • Soluble in DMSO; ultrasonic assistance may be required.
  • Store protected from light at 4°C; in solution keep at -80°C for long-term storage.
  • Available in small research quantities, such as 5 mg, for assay development.