Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Avenanthramide D | 115610-36-1 | MFCD13554294 | 99.9% | 283.28 g/mol | C16H13NO4 | 1 ML
SDP

Catalog No. 5000228965
Encompass_Preferred

Avenanthramide D is a phenolic amide found in oats (Avena sativa) and used as a research reagent to study inflammation and antioxidant activity. The compound is provided as a preprepared 10 mM solution in DMSO (1 mL) or as solid material and is supplied with analytical data and safety documentation.

  • Used for research into inflammation and oxidative stress.
  • Reported purity 99.87%.
  • Supplied as a 10 mM solution in DMSO, 1 mL, and as solids in multiple sizes.
  • Analytical data available: COA, HNMR, RP-HPLC, MS, and SDS.
  • Molecular formula C16H13NO4; molecular weight 283.28 g/mol.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-28965 Supplier Medchemexpress LLC Supplier No. HY11869310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avenanthramide D is a phenolic amide found in oats (Avena sativa) and used as a research reagent to study inflammation and antioxidant activity. The compound is provided as a preprepared 10 mM solution in DMSO (1 mL) or as solid material and is supplied with analytical data and safety documentation.

  • Used for research into inflammation and oxidative stress.
  • Reported purity 99.87%.
  • Supplied as a 10 mM solution in DMSO, 1 mL, and as solids in multiple sizes.
  • Analytical data available: COA, HNMR, RP-HPLC, MS, and SDS.
  • Molecular formula C16H13NO4; molecular weight 283.28 g/mol.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.